Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Sequence of this protein is as follows: MAWKSGGASH SELIHNLRKN GIIKTDKVFE VMLATDRSHY AKCNPYMDSP QSIGFQATIS APHMHAYALE LLFDQLHEGA KALDVGSGSG ILTACFARMV GCTGKVIGID HIKELVDDSV NNVRKDDPTL LSSGRVQLVV GDGRMGYAEE APYDAIHVGA AAPVVPQALI DQLKPGGRLI LPVGPAGGNQ MLEQYDKLQD GSIKMKPLMG VIYVPLTDKE KQWSRDEL
Three classes of protein carboxyl methyltransferases, distinguished by their methyl-acceptor substrate specificity, have been found in prokaryotic and eukaryotic cells. The type II enzyme catalyzes the transfer of a methyl group from S-adenosyl-L-methionine to the free carboxyl groups of D-aspartyl and L-isoaspartyl residues. These methyl-accepting residues result from the spontaneous deamidation, isomerization, and racemization of normal L-aspartyl and L-asparaginyl residues and represent sites of covalent damage to aging proteins PCMT1 (EC 2.1.1.77) is a protein repair enzyme that initiates the conversion of abnormal D-aspartyl and L-isoaspartyl residues to the normal L-aspartyl form.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: L-isoaspartyl protein carboxyl methyltransferase; PIMT; Protein L-isoaspartyl/D-aspartyl methyltransferase; Protein-beta-aspartate methyltransferase; Protein-L-isoaspartate(D-aspartate) O-methyltransferase
Gene Aliases: PCM; PCMT1; PIMT
UniProt ID: (Human) P22061, (Rat) P22062
Entrez Gene ID: (Human) 5110, (Rat) 25604
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support