Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MPKGRQKVPH LDAPLGLPTC LWLELAGLFL LVPWVMGLAG TGGPDGQGTG GPSWAVHLES LEGDGEEETL EQQADALAQA AGLVNAGRIG ELQGHYLFVQ PAGHRPALEV EAIRQQVEAV LAGHEAVRWH SEQRLLRRAK RSVHFNDPKY PQQWHLNNRR SPGRDINVTG VWERNVTGRG VTVVVVDDGV EHTIQDIAPN YSPEGSYDLN SNDPDPMPHP DVENGNHHGT RCAGEIAAVP NNSFCAVGVA YGSRIAGIRV LDGPLTDSME AVAFNKHYQI NDIYSCSWGP DDDGKTVDGP HQLGKAALQH GVIAGRQGFG SIFVVASGNG GQHNDNCNYD GYANSIYTVT IGAVDEEGRM PFYAEECASM LAVTFSGGDK MLRSIVTTDW DLQKGTGCTE GHTGTSAAAP LAAGMIALML QVRPCLTWRD VQHIIVFTAT RYEDRRAEWV TNEAGFSHSH QHGFGLLNAW RLVNAAKIWT SVPYLASYVS PVLKENKAIP QSPRSLEVLW NVSRMDLEMS GLKTLEHVAV TVSITHPRRG SLELKLFCPS GMMSLIGAPR SMDSWLCVEC SRHQGQTKAV RECHEWKIPA R
PCSK7 is likely to represent a ubiquitous endoprotease activity within constitutive secretory pathways and capable of cleavage at the RXXX[KR]R consensus motif.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hPC8; Lymphoma proprotein convertase; PC7; PC8; Prohormone convertase 7; prohormone convertase PC7; Proprotein convertase 7; Proprotein convertase 8; proprotein convertase PC7; Proprotein convertase subtilisin/kexin type 7; proprotein convertase subtilisin/kexin type 7 precursor variant 2; Subtilisin/kexin-like protease PC7
Gene Aliases: LPC; PC7; PC8; PCSK7; SPC7
UniProt ID: (Human) Q16549
Entrez Gene ID: (Human) 9159
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support