Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSSPQSRNGK DSKERVSRKM SIQEYELIHK EKEDENCLRK YRRQCMQDMH QKLSFGPRYG FVYELETGKQ FLETIEKELK ITTIVVHIYE DGIKGCDALN SSLTCLAAEY PIVKFCKIKA SNTGAGDRFS LDVLPTLLIY KGGELISNFI SVAEQFAEEF FAGDVESFLN EYGLLPEREV HVLEHTKIEE EDVE
PDC encodes a phosphoprotein, which is located in the outer and inner segments of the rod cells in the retina. This protein may participate in the regulation of visual phototransduction or in the integration of photoreceptor metabolism. It modulates the phototransduction cascade by interacting with the beta and gamma subunits of the retinal G-protein transducin. PDC is a potential candidate gene for retinitis pigmentosa and Usher syndrome type II. Alternatively spliced transcript variants encoding different isoforms have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 33 kDa phototransducing protein; G beta gamma binding protein; PHD; Phosducin; phosducin-like orphan protein; Protein MEKA
Gene Aliases: MEKA; PDC; PHD; PhLOP; PhLP
UniProt ID: (Human) P20941
Entrez Gene ID: (Human) 5132
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support