Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: MANLEESFPR GGTRKIHKPE KAFQQSVEQD NLFDISTEEG STKRKKSQKG PAKTKKLKIE KRESSKSARE KFEILSVESL CEGMRILGCV KEVNELELVI SLPNGLQGFV
PDCD11 is a NF-kappa-B (NFKB1)-binding protein that colocalizes with U3 RNA in the nucleolus and is required for rRNA maturation and generation of 18S rRNA.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: apoptosis-linked gene 4; NF-kappa-B-binding protein; NFBP; Programmed cell death protein 11; Protein RRP5 homolog
Gene Aliases: ALG-4; ALG4; KIAA0185; NFBP; PDCD11; RRP5
UniProt ID: (Human) Q14690
Entrez Gene ID: (Human) 22984
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support