Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAAVLKPVL LGLRDAPVHG SPTGPGAWTA SKLGGIPDAL PTVAAPRPVC QRCGQPLALV VQVYCPLEGS PFHRLLHVFA CACPGCSTGG ARSWKVFRSQ CLQVPEREAQ DAQKQGNSLA AEDWCEGADD WGSDTEEGPS PQFTLDFGND ASSAKDVDWT ARLQDLRLQD AVLGAAHPVP PGLPLFLPYY ICVADEDDYR DFVNLDHAHS LLRDYQQREG IAMDQLLSQS LPNDGDEKYE KTIIKSGDQT FYKFMKRIAA CQEQILRYSW SGEPLFLTCP TSEVTELPAC SQCGGQRIFE FQLMPALVSM LKSANLGLSV EFGTILVYTC EKSCWPPNHQ TPMEEFCIIQ EDPDELLFK (1-358 aa encoded by BC006146 )
Over expression of PDCD2L attenutates TNF-alpha release in Daudi cells. PDCD2L over-expression restrained proliferation of HEK293T cells. DNA/flow cytometry analysis showed that the over-expression of PDCD2L severely delays cell cycle progression at S phase. These all suggest that PDCD2L might play a role in apoptosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC13096; programmed cell death 2-like; Programmed cell death protein 2-like
Gene Aliases: 6030457N17Rik; PDCD2L
UniProt ID: (Human) Q9BRP1, (Mouse) Q8C5N5
Entrez Gene ID: (Human) 84306, (Mouse) 68079
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support