Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MFLCQGFRKY LPEHLNDLKK ENFSLKLRIY FLEERMQQKY EASREDIYRR NTELKVEVES LKRELQDKKQ HLDKTWADVE NLNSQNEAEL RRQFEERQQE TEHVYELLEN KMQLLQEESR LAKNEAARMA ALVEAEKECN LELSEKLKGV TKNWEDVPGD QVKPDQYTEA LAQRDKI
The protein encoded by this gene serves to anchor phosphodiesterase 4D to the Golgi/centrosome region of the cell. Defects in this gene may be a cause of myeloproliferative disorder (MBD) associated with eosinophilia. Several transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cardiomyopathy-associated protein 2; Myomegalin; myomegalin/phosphodiesterase 4D interacting protein variant 8; Phosphodiesterase 4D-interacting protein
Gene Aliases: CMYA2; KIAA0454; KIAA0477; MMGL; PDE4DIP
UniProt ID: (Human) Q5VU43
Entrez Gene ID: (Human) 9659
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support