Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • PDE5 Antibodies

          Fabgennix

          PDE5A Polyclonal Antibody, Biotin

          View all (22) PDE5 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite PDE5A Polyclonal Antibody, Biotin

          Product Details

          PD5A-BIOTIN

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Immunohistochemistry (IHC)

          1:50-1:250
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:250
          -

          ELISA (ELISA)

          1:10,000
          -

          Immunoprecipitation (IP)

          1:50-1:250
          -

          Immunomicroscopy (IM)

          1:50-1:200
          -
          Product Specifications

          Species Reactivity

          Chicken, Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide common to all PDE5A variants taken within amino acid region 825-875 on human PDE5A protein.
          View immunogen

          Conjugate

          Biotin Biotin Biotin

          Form

          Liquid

          Concentration

          0.5-1.5 mg/mL

          Purification

          Affinity chromatography

          Storage buffer

          proprietary buffer, pH 7.4-7.8, with 0.5% BSA, 30% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Target Information

          The cyclic monophosphate nucleotides (cyclic adenosine monophosphate [cAMP] and cyclic guanosine monophosphate [cGMP] are found ubiquitously in mammalian cells and act as second messenger transducers to effect the intracellular actions of a variety of G protein coupled receptors (GPCRs) for hormones, cytokines, and neurotransmitters. Cyclic nucleotides are important intracellular second messenger which play important role in a variety of signal transduction process.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: CGB-PDE; cGMP-binding cGMP specific phosphodiesterase 5A2; cGMP-binding cGMP-specific 3',5'-cyclic nucleotide phosphodiesterase; cGMP-binding cGMP-specific phosphodiesterase; cGMP-binding/cGMP-specific phosphodiesterase; cGMP-specific 3',5'-cyclic phosphodiesterase; cGMP-specific phosphodiesterase PDE5A2; cGMP-specific phosphodiesterase type 5A; phosphodiesterase 5A, cGMP-specific; phosphodiesterase isozyme 5; phosphodiesterase type 5

          View more View less

          Gene Aliases: CGB-PDE; Cgbpde; CN5A; Cn5n; PDE5; PDE5A; Pde5a1; PDE5A2

          View more View less

          UniProt ID: (Human) O76074, (Rat) O54735, (Mouse) Q8CG03

          View more View less

          Entrez Gene ID: (Human) 8654, (Rat) 171115, (Mouse) 242202

          View more View less

          Function(s)
          3',5'-cyclic-nucleotide phosphodiesterase activity cGMP binding metal ion binding 3',5'-cyclic-GMP phosphodiesterase activity cyclic-nucleotide phosphodiesterase activity nucleotide binding catalytic activity phosphoric diester hydrolase activity hydrolase activity
          Process(es)
          response to hypoxia regulation of the force of heart contraction positive regulation of chronic inflammatory response signal transduction nervous system development short-term memory positive regulation of cardiac muscle hypertrophy regulation of cGMP metabolic process response to lipopolysaccharide response to testosterone negative regulation of T cell proliferation vasodilation positive regulation of apoptotic process positive regulation of MAP kinase activity positive regulation of vasoconstriction cGMP metabolic process cGMP catabolic process negative regulation of cardiac muscle contraction relaxation of cardiac muscle positive regulation of oocyte development metabolic process positive regulation of G-protein coupled receptor protein signaling pathway
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0