Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
OriGene
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
The second messengers cAMP and cGMP are key regulatory molecules that are involved in a wide variety of signal transduction pathways. Levels of cAMP and cGMP are regulated by their rate of synthesis by nucleotide cyclases and by their rate of hydrolysis by cyclic nucleotide phosphodiesterases (PDEs). PDEs form a superfamily of enzymes that catalyze the conversion of 3-prime, 5-prime-cyclic nucleotides to the corresponding nucleoside 5-prime-monophosphates. PDE6 is the effector enzyme in the G protein-mediated signal transduction cascade in the visual system. There are five different subunits consisting of rod and cone specific catalytic subunits: alpha' (Cone), alpha (Rod), and beta (Rod), the inhibitory subunit gamma, and subunit delta of unknown function (which likely interacts with many other proteins besides the PDE6 family). The catalytic core of the PDE6 system is comprised of alpha'/alpha' homodimers in the cone and alpha/beta heterodimers in the rod. The C-terminus of both the catalytic and inhibitory subunits is modified by methylation, myristyolation and prenylation which have been shown to be critical for proper complex assembly and membrane association.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GMP-PDE gamma; phosphodiesterase 6G, cGMP-specific, rod, gamma; Retinal rod rhodopsin-sensitive cGMP 3',5'-cyclic phosphodiesterase subunit gamma; rod cG-PDE G
Gene Aliases: PDE6G; PDEG; RP57
UniProt ID: (Human) P18545
Entrez Gene ID: (Human) 5148
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support