Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSDNTTLPA PASNQGPTTP RKGPPKFKQR QTRQFKSKPP KKGVKGFGDD IPGMEGLGTD ITVICPWEAF SHLELHELAQ FGII (1-83 aa encoded by BC093738)
Participates in processes of transmission and amplification of the visual signal. cGMP-PDEs are the effector molecules in G-protein-mediated phototransduction in vertebrate rods and cones.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cone phosphodiesterase 6 gamma; cone-like cGMP-phosphodiesterase 6 gamma subunit type 2; GMP-PDE gamma; phosphodiesterase 6G, cGMP-specific, rod, gamma; phosphodiesterase 6H, cGMP-specific, cone, gamma; Retinal cone rhodopsin-sensitive cGMP 3',5'-cyclic phosphodiesterase subunit gamma
Gene Aliases: A930033D18Rik; ACHM6; Pde6g; PDE6H; PDEgamma; RCD3
UniProt ID: (Human) Q13956, (Rat) P61250, (Mouse) P61249
Entrez Gene ID: (Human) 5149, (Rat) 114248, (Mouse) 78600
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support