Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
The PDHX gene encodes component X of the pyruvate dehydrogenase (PDH) complex. For a detailed description of the pyruvate dehydrogenase complex. The mammalian PDH complex differs from that in E. coli and from the other mammalian alpha-keto acid dehydrogenases by the presence of a 53-kDa protein called protein X. Component X binds to the E3 component of the PDH complex (Robinson et al., 1990 ; Aral et al., 1997 ).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: component X; Dihydrolipoamide dehydrogenase-binding protein of pyruvate dehydrogenase complex; dihydrolipoamide dehydrogenase-binding protein of pyruvate dehydrogenase complex (E3-binding protein) (proX); E3-binding protein; E3BP; GSF; IDX-1; IPF-1; IPF1; IUF-1; lipoyl-containin; Lipoyl-containing pyruvate dehydrogenase complex component X; PDX-1; proX; pyruvate dehydrogenase; pyruvate dehydrogenase complex, component X; pyruvate dehydrogenase complex, E3-binding protein subunit; pyruvate dehydrogenase complex, lipoyl-containing component X; Pyruvate dehydrogenase protein X component, mitochondrial; STF-1
Gene Aliases: Ac1164; AI481367; DLDBP; E3BP; OPDX; PDHX; PDX1; proX
UniProt ID: (Human) O00330, (Mouse) Q8BKZ9
Entrez Gene ID: (Human) 8050, (Rat) 311254, (Mouse) 27402
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support