Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: VAGAEGPDED SSNRENAIED EEEEEEEDDD EEEDDLEVKE ENGVLVLNDA NFDNFVADKD TVLLEFYAPW CGHCKQFAPE YEKIANILKD KDPPIPVAKI DATSASVLAS RFDVSGYPTI KILKKGQAVD YEGSRTQEEI VAKVREVSQP DWTPPPEVTL VLTKENFDEV VNDADIILVE FYAPWCGHCK KLAPEYEKAA KELSKRSPPI PLAKVDATAE TDLAKRFDVS GYPTLKIFRK GRPYDYNGPR EKYGIVDYMI EQSGPPSKEI LTLKQVQEFL KDGDDVIIIG VFKGESDPAY QQYQDAANNL REDYKFHHTF STEIAKFLKV SQGQLVVMQP EKFQSKYEPR SHMMDVQGST QDSAIKDFVL KYALPLVGHR KVSNDAKRYT RRPLVVVYYS VDFSFDYRAA TQFWRSKVLE VAKDFPEYTF AIADEEDYAG EVKDLGLSES GEDVNAAILD ESGKKFAMEP EEFDSDTLRE FVTAFKKGKL KPVIKSQPVP KNNKGPVKVV VGKTFDSIVM DPKKDVLIEF YAPWCGHCKQ LEPVYNSLAK KYKGQKGLVI AKMDATANDV PSDRYKVEGF PTIYFAPSGD KKNPVKFEGG DRDLEHLSKF IEEHATKLSR TKEEL
ERp72 is an endoplasmic reticulum luminal protein that is both a stress protein and a member of the protein disulfide isomerase family of proteins. Sequence analysis of ERp72 shows that it shares sequence identity with PDI (protein disulphide isomerase) at three discrete regions, as well as containing the ER retention signal KEEL at its C-terminus. ERp72 is shown to have three copies of sequence believed to be CGHC-containing active sites, which are also found in PDI. These CGHC sites are critical sites that are involved in the protein disulphide isomerization process. These results suggest that ERp72 is a newly described member of the family of CGHC-containing proteins that functions as a molecular chaperone.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EC 5.3.4.1; Endoplasmic reticulum resident protein 70; Endoplasmic reticulum resident protein 72; ER protein 70; ER protein 72; protein disu; protein disulfide isomerase family A, member 4; protein disulfide isomerase related protein (calcium-binding protein, intestinal-related); protein disulfide isomerase-associated 4; Protein disulfide-isomerase A4; Protein ERp-72; testicular tissue protein Li 137
Gene Aliases: ERp-72; ERP70; ERP72; PDIA4
UniProt ID: (Human) P13667
Entrez Gene ID: (Human) 9601
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support