Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:64,000.
Pyridoxal 5-prime-phosphate (PLP) is the active form of vitamin B6 that acts as a coenzyme in maintaining biochemical homeostasis. The preferred degradation route from PLP to 4-pyridoxic acid involves the dephosphorylation of PLP by PDXP. Pyridoxal 5-prime-phosphate (PLP) is the active form of vitamin B6 that acts as a coenzyme in maintaining biochemical homeostasis. The preferred degradation route from PLP to 4-pyridoxic acid involves the dephosphorylation of PLP by PDXP (Jang et al., 2003 ).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: chronophilin; Chronophin; FLJ32703; PLP phosphatase; PLP-phosophatase; pyridoxal (pyridoxine, vitamin B6) phosphatase; Pyridoxal 5'-phosphate phosphatase; pyridoxal phosphatase; Pyridoxal phosphate phosphatase; reg I binding protein I; reg I-binding protein 1; testicular secretory protein Li 36
Gene Aliases: 1600027H05Rik; AB041662; CIN; dJ37E16.5; PDXP; PLP; PLPP; Rbp1
UniProt ID: (Human) Q96GD0, (Mouse) P60487
Entrez Gene ID: (Human) 57026, (Mouse) 57028, (Rat) 727679
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support