Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAWQGLVLAA CLLMFPSTTA DCLSRCSLCA VKTQDGPKPI NPLICSLQCQ AALLPSEEWE RCQSFLSFFT PSTLGLNDKE DLGSKSVGEG PYSELAKLSG SFLKELEKSK FLPSISTKEN TLSKSLEEKL RGLSDGFREG AESELMRDAQ LNDGAMETGT LYLAEEDPKE QVKRYGGFLR KYPKRSSEVA GEGDGDSMGH EDLYKRYGGF LRRIRPKLKW DNQKRYGGFL RRQFKVVTRS QEDPNAYSGE LFDA
expression in hippocampus increases in response to traumatic brain injury; may play a role in regulatio of neurotransmitter release.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Beta-neoendorphin-dynorphin; leu-enkephalin; leumorphin; neoendorphin-dynorphin-enkephalin prepropeptide; Preprodynorphin; preproenkephalin B; Proenkephalin-B; rimorphin
Gene Aliases: ADCA; PDYN; PENKB; SCA23
UniProt ID: (Human) P01213
Entrez Gene ID: (Human) 5173
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support