Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
Immunogen Sequence: APGGPYGPPA GGGPYGHPNP GMFPSGTPGG PYGGAAPGGP YGQPPPSSYG AQQPGLYGQG GAPPNVDPEA YSWFQSVDSD HSGYISMKEL KQALVNCNWS S
PEF1 is a Ca(2+)-binding protein that belongs to the penta-EF-hand (PEF) protein family, which includes the calpain small subunit (CAPNS1), sorcin (SRI), grancalcin (GCA), and ALG2. PEF1 is a Ca(2+)-binding protein that belongs to the penta-EF-hand (PEF) protein family, which includes the calpain small subunit (CAPNS1), sorcin (SRI), grancalcin (GCA), and ALG2 (PDCD6) (Kitaura et al., 2001 ).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: PEF protein with a long N-terminal hydrophobic domain; Peflin; penta-EF hand domain containing 1; Penta-EF hand domain-containing protein 1
Gene Aliases: 2600002E23Rik; ABP32; PEF1; PEF1A; Peflin; UNQ1845/PRO3573
UniProt ID: (Human) Q9UBV8, (Mouse) Q8BFY6, (Rat) Q641Z8
Entrez Gene ID: (Human) 553115, (Mouse) 67898, (Rat) 297900
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support