Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Allows the formation of correctly charged Gln-tRNA(Gln) through the transamidation of misacylated Glu-tRNA(Gln) in the mitochondria. The reaction takes place in the presence of glutamine and ATP through an activated gamma-phospho-Glu-tRNA(Gln). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cytochrome c oxidase assembly factor PET112 homolog; cytochrome oxidase assembly factor PET112 homolog; Glu-AdT subunit B; Glutamyl-tRNA(Gln) amidotransferase subunit B, mitochondrial; glutamyl-tRNA(Gln) amidotransferase, subunit B; PET112 homolog
Gene Aliases: GATB; HSPC199; PET112; PET112L
UniProt ID: (Human) O75879
Entrez Gene ID: (Human) 5188
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support