Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MDAFTRFTNQ TQGRDRLFRA TQYTCMLLRY LLEPKAGKEK VVMKLKKLES SVSTGRKWFR LGNVVHAIQA TEQSIHATDL VPRLCLTLAN LNR
PEX11A may be involved in peroxisomal proliferation and may regulate peroxisomes division. It may mediate binding of coatomer proteins to the peroxisomal membrane.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 28 kDa peroxisomal integral membrane protein; HsPEX11p; Peroxin-11A; Peroxisomal biogenesis factor 11A; Peroxisomal membrane protein 11A; PEX11-alpha; PMP28; Protein PEX11 homolog alpha
Gene Aliases: hsPEX11p; PEX11; PEX11-ALPHA; PEX11A; PMP28
UniProt ID: (Human) O75192
Entrez Gene ID: (Human) 8800
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support