Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MKWMVVVLVC LQLLEAAVVK VPLKKFKSIR ETMKEKGLLG EFLRTHKYDP AWKYRFGDLS VTYEPMAYMD AAYFGEISIG TPPQNFLVLF DTGSSNLWVP SVYCQSQACT SHSRFNPSES STYSTNGQTF SLQYGSGSLT GFFGYDTLTV QSIQVPNQEF GLSENEPGTN FVYAQFDGIM GLAYPALSVD EATTAMQGMV QEGALTSPVF SVYLSNQQGS SGGAVVFGGV DSSLYTGQIY WAPVTQELYW QIGIEEFLIG GQASGWCSEG CQAIVDTGTS LLTVPQQYMS ALLQATGAQE DEYGQFLVNC NSIQNLPSLT FIINGVEFPL PPSSYILSNN GYCTVGVEPT YLSSQNGQPL WILGDVFLRS YYSVYDLGNN RVGFATAA
Pepsin is one of the main proteolytic enzymes secreted by the gastric mucosa. Pepsin consists of a single polypeptide chain and arises from its precursor, pepsinogen, by removal of a 41 amino acid segment from the N-terminus. Pepsinogen is synthesized in the stomach lining, and hydrochloric acid, also produced by the gastric mucosa, is necessary to convert the inactive enzyme and to maintain the optimum acidity (pH 1-3) for pepsin function. Pepsin is particularly effective in cleaving peptide bonds involving aromatic amino acids. Pepsin shows extremely broad specificity, and although bonds involving phenylalanine and leucine are preferred, many others are also cleaved to some extent. The amino acid composition of Pepsin C differs from those of pepsinogen and pepsin especially in the content of basic amino acids, glutamic acid, aspartic acid, leucine and isoleucine.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Gastricsin; pep carboxylase; pepcase; pepsin C; Pepsinogen C; pepsinogen group II; preprogastricsin; progastricsin (pepsinogen C)
Gene Aliases: PEPC; PGC; PGII
UniProt ID: (Human) P20142
Entrez Gene ID: (Human) 5225
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support