Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: IKRGVLKEI YDKDGELSIS NEEDSLENGQ SLSSSQLSLP ALSEMEPVPM PRDPCSYEVL QPSDIMDGPD PGAPVKLPCL PVKLSPPLPP KKVMICMPVG GPDLSLVSYT AQKSGQQGVA QHHHTVLPSQ IQHQLQYGSH GQHLPSTTGS LPMHPSGCRM IDELNKTLAM TMQRLESSEQ RVPCSTSYHS SGLHSGDGVT KAGPMGLPEI RQVPTVVIEC DDNKENVPHE SDYEDSSCLY TREEEEEEED EDDDSSLYTS SLAMKVCRKD SLAI (154-426 aa encoded by B C101526)
PHACTR1 plays a role in cell motility and in the formation of tubules by endothelial cells. PHACTR1 also binds actin monomers (G actin) and plays a role in the reorganization of the actin cytoskeleton and in formation of actin stress fibers. Further, PHACTR1 regulates PPP1CA activity and is required for normal cell survival.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC126575; MGC126577; Phosphatase and actin regulator 1; RPEL repeat containing 1
Gene Aliases: 9630030F18Rik; dJ257A7.2; KIAA1733; PHACTR1; RPEL; RPEL1
UniProt ID: (Human) Q9C0D0, (Rat) P62024, (Mouse) Q2M3X8
Entrez Gene ID: (Human) 221692, (Rat) 306844, (Mouse) 218194
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support