Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • PHKA2 Antibodies

          Abnova

          PHKA2 Monoclonal Antibody (1D4)

          View all (7) PHKA2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite PHKA2 Monoclonal Antibody (1D4)

          • Antibody Testing Data (1)
          PHKA2 Antibody in ELISA (ELISA)
          Group 53 Created with Sketch.
          PHKA2 Antibody in ELISA (ELISA)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PHKA2 Antibody (H00005256-M05) in ELISA

          Detection limit for recombinant GST tagged PHKA2 is 1 ng/mL as a capture antibody. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PHKA2 Antibody in ELISA (ELISA)

          Product Details

          H00005256-M05

          Applications
          Tested Dilution
          Publications

          ELISA (ELISA)

          10 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG2b, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          1D4

          Immunogen

          PHKA2 (AAH14036, 1 a.a. approximately 1235 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Wet ice

          Product Specific Information

          Product may be used with Sandwich ELISA (Recombinant protein).

          Immunogen sequence: MRSRSNSGVR LDGYARLVQQ TILCYQNPVT GLLSASHEQK DAWVRDNIYS ILAVWGLGMA YRKNADRDED KAKAYELEQN VVKLMRGLLQ CMMRQVAKVE KFKHTQSTKD SLHAKYNTAT CGTVVGDDQW GHLQVDATSL FLLFLAQMTA SGLRIIFTLD EVAFIQNLVF YIEAAYKVAD YGMWERGDKT NQGIPELNAS SVGMAKAALE AIDELDLFGA HGGRKSVIHV LPDEVEHCQS ILFSMLPRAS TSKEIDAGLL SIISFPAFAV EDVNLVNVTK NEIISKLQGR YGCCRFLRDG YKTPREDPNR LHYDPAELKL FENIECEWPV FWTYFIIDGV FSGDAVQVQE YREALEGILI RGKNGIRLVP ELYAVPPNKV DEEYKNPHTV DRVPMGKVPH LWGQSLYILS SLLAEGFLAA GEIDPLNRRF STSVKPDVVV QVTVLAENNH IKDLLRKHGV NVQSIADIHP IQVQPGRILS HIYAKLGRNK NMNLSGRPYR HIGVLGTSKL YVIRNQIFTF TPQFTDEHHF YLALDNEMIV EMLRIELAYL CTCWRMTGRP TLTFPISRTM LTNDGSDIHS AVLSTIRKLE DGYFGGARVK LGNLSEFLTT SFYTYLTFLD PDCDEKLFDN ASEGTFSPDS DSDLVGYLED TCNQESQDEL DHYINHLLQS TSLRSYLPPL CKNTEDRHVF SAIHSTRDIL SVMAKAKGLE VPFVPMTLPT KVLSAHRKSL NLVDSPQPLL EKVPESDFQW PRDDHSDVDC EKLVEQLKDC SNLQDQADIL YILYVIKGPS WDTNLSGQHG VTVQNLLGEL YGKAGLNQEW GLIRYISGLL RKKVEVLAEA CTDLLSHQKQ LTVGLPPEPR EKIISAPLPP EELTKLIYEA SGQDISIAVL TQEIVVYLAM YVRAQPSLFV EMLRLRIGLI IQVMATELAR SLNCSGEEAS ESLMNLSPFD MKNLLHHILS GKEFGVERSV RPIHSSTSSP TISIHEVGHT GVTKTERSGI NRLRSEMKQM TRRFSADEQF FSVGQAASSS AHSSKSARSS TPSSPTGTSS SDSGGHHIGW GERQGQWLRR RRLDGAINRV PVGFYQRVWK ILQKCHGLSI DGYVLPSSTT REMTPHEIKF AVHVESVLNR VPQPEYRQLL VEAIMVLTLL SDTEMTSIGG IIHVDQIVQM ASQLFLQDQV SIGAMDTLEK DQATGICHFF YDSAPSGAYG TMTYLTRAVA SYLQELLPNS GCQMQ

          Target Information

          Phosphorylase b kinase catalyzes the phosphorylation of serine in certain substrates, including troponin I. The alpha chain may bind calmodulin.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: phosphorylase b kinase regulatory subunit alpha liver isoform; Phosphorylase b kinase regulatory subunit alpha, liver isoform; phosphorylase kinase; Phosphorylase kinase alpha L subunit; phosphorylase kinase alpha-subunit; phosphorylase kinase, alpha 2 (liver); RP3-499B10.2

          View more View less

          Gene Aliases: GSD9A; PHK; PHKA2; PHKLA; PYK; PYKL; XLG; XLG2

          View more View less

          UniProt ID: (Human) P46019

          View more View less

          Entrez Gene ID: (Human) 5256

          View more View less

          Function(s)
          phosphorylase kinase activity calmodulin binding
          Process(es)
          carbohydrate metabolic process glycogen catabolic process generation of precursor metabolites and energy cellular protein modification process protein phosphorylation
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0