Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
OriGene
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
PHYH2 catalyzes a carbon-carbon cleavage reaction; cleaves a 2-hydroxy-3-methylacyl-CoA into formyl-CoA and a 2-methyl-branched fatty aldehyde.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1600020H07Rik; 2-HPCL; HPCL; HPCL2; Lupus nephritis-associated peptide 1; OTTHUMP00000019131; OTTHUMP00000019132; OTTHUMP00000179083; OTTHUMP00000216226; PhyH; PHYH2; Phytanic acid oxidase; phytanoil-CoA alpha hydroxylase; phytanoyl-CoA 2 oxoglutarate dioxygenase; Phytanoyl-CoA alpha-hydroxylase; Phytanoyl-CoA dioxygenase, peroxisomal
Gene Aliases: AI256161; AI265699; LN1; LNAP1; PAHX; PHYH; PHYH1; RD
UniProt ID: (Human) O14832, (Mouse) O35386
Entrez Gene ID: (Human) 5264, (Mouse) 16922
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support