Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: RLQGNPLGEA SPDAPSSPVA ALIPEMPRLF LTSDLDSFPV TPQGCSVTLA
Sequence homology: Human: 100%; Mouse: 86%; Rat: 100%
The protein encoded by this gene contains a leucine-rich repeat and a death domain. This protein has been shown to interact with other death domain proteins, such as Fas (TNFRSF6)-associated via death domain (FADD) and MAP-kinase activating death domain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp434D229; Leucine-rich repeat and death domain-containing protein; leucine-rich repeats and death domain containing; MGC16925; p53-induced death domain-containing protein 1; p53-induced protein with a death domain
Gene Aliases: LRDD; PIDD; PIDD1
UniProt ID: (Human) Q9HB75
Entrez Gene ID: (Human) 55367
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support