Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: GGLDGDMFNY YKMLMLQGLI AARKHMDKVV QIVEIMQQGS QLPCFHGSST IRNLKERFHM SMTEEQLQLL VEQMVDGSMR SITTKLYDGF QYLTNGIM
PI4KB and PI4KA are the two human type III PI4 kinases, which share sequence similarly with PI3 kinases and phosphorylate the 4 position of phosphotidyl inositol. Transporter for alanine, serine, cysteine, and threonine. Exhibits sodium dependence. Target protein phosphorylates phosphatidylinositol (PtdIns) at the D4 position of the inositol ring; may play a role in neuronal differentiation and maturation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: NPIK; phosphatidylinositol 4-kinase; Phosphatidylinositol 4-kinase beta; phosphatidylinositol 4-kinase, catalytic, beta; phosphatidylinositol 4-kinase, wortmannin-sensitive; PI4K-beta; PI4K92; PI4KIII; PtdIns 4-kinase beta; type III phosphatidylinositol 4-kinase beta
Gene Aliases: NPIK; PI4K-BETA; PI4K92; PI4KB; PI4KBETA; PI4KIIIBETA; PIK4CB
UniProt ID: (Human) Q9UBF8
Entrez Gene ID: (Human) 5298
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support