Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen sequence: MSSNCTSTTA VAVAPLSASK TKTKKKHFVC QKVKLFRASE PILSVLMWGV NHTINELSNV PVPVMLMPDD FKAYSKIKVD NHLFNKENLP SRFKFKEYCP MVFRNLRERF GIDDQDYQNS VTRSAPINSD SQGRCGTRFL TTYDRRFVIK TVSSEDVAEM HNILKKYHQF IVECHGNTLL PQFLGMYRLT VDGVETYMVV TRNVFSHRLT VHRKYDLKGS TVAREASDKE KAKDLPTFKD NDFLNEGQKL HVGEESKKNF LEKLKRDVEF; Positive Samples: BT-474, HepG2, A-549; Cellular Location: Cell membrane, Cytoplasm, Endoplasmic reticulum membrane, Nucleus, Peripheral membrane protein
The protein encoded by this gene catalyzes the phosphorylation of phosphatidylinositol-5-phosphate on the fourth hydroxyl of the myo-inositol ring to form phosphatidylinositol-5,4-bisphosphate. This gene is a member of the phosphatidylinositol-5-phosphate 4-kinase family. The encoded protein sequence does not show similarity to other kinases, but the protein does exhibit kinase activity. Additionally, the encoded protein interacts with p55 TNF receptor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1-phosphatidylinositol 5-phosphate 4-kinase 2-beta; 1-phosphatidylinositol-4-phosphate kinase; 1-phosphatidylinositol-5-phosphate 4-kinase 2-beta; Diphosphoinositide kinase 2-beta; Phosphatidylinositol 5-phosphate 4-kinase type II beta; Phosphatidylinositol 5-phosphate 4-kinase type-2 beta; phosphatidylinositol-4-phosphate 5-kinase, type II, beta; phosphatidylinositol-5-phosphate 4-kinase type-2 beta; phosphatidylinositol-5-phosphate 4-kinase, type II, beta; PI(5)P 4-kinase type II beta; PIP4KII-beta; PTDINS(4)P-5-kinase; PtdIns(5)P-4-kinase isoform 2-beta
Gene Aliases: PI5P4KB; PIP4K2B; PIP5K2B; PIP5KIIB; PIP5KIIbeta; PIP5P4KB
UniProt ID: (Human) P78356
Entrez Gene ID: (Human) 8396
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support