Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MLERPPALAM PMPTEGTPPP LSGTPIPVPA YFRHAEPGFS LKRPRGLSRS LPPPPPAKGS IPISRLFPPR TPGWHQLQPR RVSFRGEASE TLQSPGYDPS RPESFFQQSF QRLSRLGHGS YGEVFKVRSK EDGRLYAVKR SMSPFRGPKD RARKLAEVGS HEKVGQHPCC VRLEQAWEEG GILYLQTELC GPSLQQHCEA WGASLPEAQV WGYLRDTLLA LAHLHSQGLV HLDVKPANIF LGPRGRCKLG DFGLLVELGT AGAGEVQEGD PRYMAPELLQ GSYGTAADVF SLGLTILEVA CNMELPHGGE GWQQLRQGYL PPEFTAGLSS ELRSVLVMML EPDPKLRATA EALLALPVLR QPRAWGVLWC MAAEALSRGW ALWQALLALL CWLWHGLAHP ASWLQPLGPP ATPPGSPPCS LLLDSSLSSN WDDDSLGPSL SPEAVLARTV GSTSTPRSRC TPRDALDLSD INSEPPRGSF PSFEPRNLLS LFEDTLDPT
PKMYT1 serine/threonine kinase is a member of a family of neural specific, zinc finger-containing DNA-binding proteins which binds to the promoter regions of proteolipid proteins of the central nervous system and plays a role in the developing nervous system. MYT1 of the oligodendrocyte lineage, along with a closely related CCHC zinc finger, is expressed in developing neurons in the mammalian central nervous system. The recombinant PKMYT1 fragments containing either the upstream 2 zinc fingers or the downstream 4 zinc fingers bound the same cis regulatory element in the PLP1 promoter in vitro and DNA binding required Zn2+, but not other divalent cations.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Membrane-associated tyrosine- and threonine-specific cdc2-inhibitory kinase; Myt1 kinase; protein phosphatase 1, regulatory subunit 126
Gene Aliases: MYT1; PKMYT1; PPP1R126
UniProt ID: (Human) Q99640
Entrez Gene ID: (Human) 9088
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support