Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • PKN1 Antibodies

          Abnova

          PKN1 Polyclonal Antibody, MaxPab™

          View all (14) PKN1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite PKN1 Polyclonal Antibody, MaxPab™

          • Antibody Testing Data (3)
          PKN1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          PKN1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PKN1 Antibody (H00005585-D01P) in WB

          Western Blot analysis of PKN1 expression in transfected 293T cell line (H00005585-T01) by PKN1 MaxPab polyclonal antibody.

          Lane 1: PKN1 transfected lysate(103.90 KDa).
          Lane 2: Non-transfected lysate.
          {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PKN1 Antibody in Western Blot (WB)
          PKN1 Antibody in Western Blot (WB)
          PKN1 Antibody in Western Blot (WB)

          Product Details

          H00005585-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          PKN1 (AAH40061.1, 1 a.a. approximately 942 a.a) full-length human protein.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          See Label

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Sequence of this protein is as follows: MASDAVQSEP RSWSLLEQLG LAGADLAAPG VQQQLELERE RLRREIRKEL KLKEGAENLR RATTDLGRSL GPVELLLRGS SRRLDLLHQQ LQELHAHVVL PDPAATHDGP QSPGAGGPTC SATNLSRVAG LEKQLAIELK VKQGAENMIQ TYSNGSTKDR KLLLTAQQML QDSKTKIDII RMQLRRALQA GQLENQAAPD DTQGSPDLGA VELRIEELRH HFRVEHAVAE GAKNVLRLLS AAKAPDRKAV SEAQEKLTES NQKLGLLREA LERRLGELPA DHPKGRLLRE ELAAASSAAF STRLAGPFPA THYSTLCKPA PLTGTLEVRV VGCRDLPETI PWNPTPSMGG PGTPDSRPPF LSRPARGLYS RSGSLSGRSS LKAEAENTSE VSTVLKLDNT VVGQTSWKPC GPNAWDQSFT LELERARELE LAVFWRDQRG LCALKFLKLE DFLDNERHEV QLDMEPQGCL VAEVTFRNPV IERIPRLRRQ KKIFSKQQGK AFQRARQMNI DVATWVRLLR RLIPNATGTG TFSPGASPGS EARTTGDISV EKLNLGTDSD SSPQKSSRDP PSSPSSLSSP IQESTAPELP SETQETPGPA LCSPLRKSPL TLEDFKFLAV LGRGHFGKVL LSEFRPSGEL FAIKALKKGD IVARDEVESL MCEKRILAAV TSAGHPFLVN LFGCFQTPEH VCFVMEYSAG GDLMLHIHSD VFSEPRAIFY SACVVLGLQF LHEHKIVYRD LKLDNLLLDT EGYVKIADFG LCKEGMGYGD RTSTFCGTPE FLAPEVLTDT SYTRAVDWWG LGVLLYEMLV GESPFPGDDE EEVFDSIVND EVRYPRFLSA EAIGIMRRLL RRNPERRLGS SERDAEDVKK QPFFRTLGWE ALLARRLPPP FVPTLSGRTD ISNFDEEFTG EAPTLSPPRD ARPLTAAEQA AFLDFDFVAG GC

          Target Information

          PKN1 (PRK1) belongs to the protein kinase C superfamily. This kinase is activated by the Rho family of small G proteins and may mediate the Rho-dependent signaling pathway. This kinase can also be activated by phospholipids. The 3-phosphoinositide dependent protein kinase-1 (PDPK1/PDK1) is reported to phosphorylate this kinase, which may mediate insulin signals to the actin cytoskeleton. The proteolytic activation of this kinase by caspase-3 or related proteases during apoptosis suggests its role in signal transduction related to apoptosis. Alternatively spliced transcript variants encoding distinct isoforms have been observed.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: PAK-1; Protease-activated kinase 1; Protein kinase C-like 1; Protein kinase C-like PKN; protein kinase C-related kinase 1; Protein kinase PKN-alpha; Protein-kinase C-related kinase 1; serine-threonine kinase N; Serine-threonine protein kinase N; serine/threonine kinase 3; serine/threonine protein kinase N; Serine/threonine-protein kinase N1

          View more View less

          Gene Aliases: DBK; F730027O18Rik; PAK-1; PAK1; PKN; PKN-ALPHA; PKN1; PRK1; PRKCL1; Stk3

          View more View less

          UniProt ID: (Human) Q16512, (Mouse) P70268

          View more View less

          Entrez Gene ID: (Human) 5585, (Mouse) 320795

          View more View less

          Function(s)
          chromatin binding protein kinase activity protein serine/threonine kinase activity protein kinase C activity protein kinase C binding protein binding ATP binding GTP-Rho binding ligand-dependent nuclear receptor transcription coactivator activity histone kinase activity (H3-T11 specific) histone binding histone deacetylase binding Rac GTPase binding androgen receptor binding nucleotide binding kinase activity transferase activity transferase activity, transferring phosphorus-containing groups
          Process(es)
          B cell homeostasis B cell apoptotic process regulation of germinal center formation regulation of immunoglobulin production renal system process transcription, DNA-templated regulation of transcription from RNA polymerase II promoter protein phosphorylation negative regulation of protein kinase activity hyperosmotic response signal transduction activation of JUN kinase activity epithelial cell migration peptidyl-serine phosphorylation negative regulation of B cell proliferation histone H3-T11 phosphorylation spleen development regulation of cell motility negative regulation of protein phosphorylation regulation of transcription, DNA-templated phosphorylation chromatin modification
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0