Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: ELTLYDKDI LGSDQLSLLL FDLRSLKCGQ PHKHTFPLNH QDSQELQVEF VLEKTQVPAS EVITNGVLVA HPCLRIQGTL RGDGTAPREE YGSRQLQLAV PGAYEKPQLL PLQPPTEPGL PPTFTFHVNP VLSSRLHVEL MEVLAAVQSG PSAELEAQTS KLGEGGILLS SLPLGQEEQC SVALGEGQEV ALSMKVEMSS GDLDLRLGFD LSDGEQEFLD R (111-330 aa encoded by BC146648)
Calcium-dependent phospholipase A2 that selectively hydrolyzes glycerophospholipids in the sn-2 position. Has higher enzyme activity for phosphatidylethanolamine than phosphatidylcholine.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3.1.1.4; cPLA2-zeta; Cytosolic phospholipase A2 zeta; Phospholipase A2 group IVF; phospholipase A2, group IVF
Gene Aliases: PLA2G4F; PLA2G4FZ
UniProt ID: (Human) Q68DD2
Entrez Gene ID: (Human) 255189
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support