Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • PLEKHA3 Antibodies

          Invitrogen

          PLEKHA3 Monoclonal Antibody (OTI5D11)

          View all (9) PLEKHA3 antibodies
          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite PLEKHA3 Monoclonal Antibody (OTI5D11)

          • Antibody Testing Data (4)
          PLEKHA3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          PLEKHA3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 4

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PLEKHA3 Antibody (MA5-25549) in ICC/IF

          Immunofluorescent analysis of PLEKHA3 in HeLa cells. Cells were probed with a PLEKHA3 monoclonal antibody (Product # MA5-25549). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PLEKHA3 Antibody in Immunocytochemistry (ICC/IF)
          PLEKHA3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PLEKHA3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PLEKHA3 Antibody in Western Blot (WB)

          Product Details

          MA5-25549

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:2,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:150
          -

          Immunocytochemistry (ICC/IF)

          1:100
          -

          Flow Cytometry (Flow)

          1:100
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG2b

          Class

          Monoclonal

          Type

          Antibody

          Clone

          OTI5D11

          Immunogen

          Full length human recombinant protein of PLEKHA3 produced in HEK293T cell
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1.1 mg/mL

          Purification

          Affinity Chromatography

          Storage buffer

          PBS, pH 7.3, with 1% BSA, 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2725045

          Target Information

          PLEKHA3 (Pleckstrin Homology Domain Containing A3) is a Protein Coding gene. Among its related pathways are Metabolism and Cholesterol and Sphingolipids transport / Transport from Golgi and ER to the apical membrane (normal and CF). Gene Ontology (GO) annotations related to this gene include phospholipid binding and 1-phosphatidylinositol binding. An important paralog of this gene is PLEKHA8.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: FAPP-1; four-phosphate-adaptor protein 1; PH domain-containing family A member 3; Phosphatidylinositol-four-phosphate adapter protein 1; phosphoinositol 4-phosphate adapter protein 1; pleckstrin homology domain containing, family A (phosphoinositide binding specific) member 3; Pleckstrin homology domain-containing family A member 3; pleckstrin homology domain-containing, family A (phosphoinositide binding specific) member 3

          View more View less

          Gene Aliases: FAPP1; PLEKHA3

          View more View less

          UniProt ID: (Human) Q9HB20

          View more View less

          Entrez Gene ID: (Human) 65977

          View more View less

          Function(s)
          phospholipid binding 1-phosphatidylinositol binding
          Process(es)
          biological_process
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-79b49c4fc5-ghzkj:80/100.66.129.252:80.
          git-commit: cca8d8ff64ba87988ed84f3fa7b2b2bee312c353
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0