Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Sequence of this protein is as follows: MRGELWLLVL VLREAARALS PQPGAGHDEG PGSGWAAKGT VRGWNRRARE SPGHVSEPDR TQLSQDLGGG TLAMDTLPDN RTRVVEDNHS YYVSRLYGPS EPHSRELWVD VAEANRSQVK IHTILSNTHR QASRVVLSFD FPFYGHPLRQ ITMATGGFIF MGDVIHRMLT ATQYVAPLMA NFNPGYSDNS TVVYFDNGTV FVVQWDHVYL QGWEDKGSFT FQAALHHDGR IVFAYKEIPM SVPEISSSQH PVKTGLSDAF MILNPSPDVP ESRRRSIFEY HRIELDPSKV TSMSAVEFTP LPTCLQHRSC DACMSSDLTF NCSWCHVLQR CSSGFDRYRQ EWMDYGCAQE AEGRMCEDFQ DEDHDSASPD TSFSPYDGDL TTTSSSLFID SLTTEDDTKL NPYAGGDGLQ NNLSPKTKGT PVHLGTIVGI VLAVLLVAAI ILAGIYINGH PTSNAALFFI ERRPHHWPAM KFRSHPDHST YAEVEPSGHE KEGFMEAEQC
Recently, using SAGE (Serial Analysis of Gene Expression) technology, St. Croix et al, have identified 46 genes, whose expression is specifically elevated in tumor-associated endothelium. Nine of these genes were prominently expressed only in tumor endothelial cells (EC), but were absent or barely detectable in normal ECs, and named as Tumor Endothelial Markers (TEMs, TEM 1-9). TEM7 (Tumor endothelial marker 7) transcripts are specifically expressed in the endothelium of colorectal cancer, primary cancers of lung, pancreas, breast, and brain. TEM7 is expressed specifically in endothelium of these cancers, whether primary or metastasis. The other six members of this family (TEM1, 3, 4, 5, 8, and 9) also show similar expression pattern in lung and brain tumors, and liver metastasis. Since most of the genes expressed differentially in tumor endothelium are also expressed during angiogenesis, these newly discovered genes might provide important resources for basic and clinical studies of human angiogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2410003I07Rik; DKFZp686F0937; FLJ36270; FLJ45632; Plexin domain-containing protein 1; TEM7 related; Tumor endothelial marker 3; Tumor endothelial marker 7
Gene Aliases: 2410003I07Rik; AI848450; PLXDC1; TEM3; TEM7
UniProt ID: (Human) Q8IUK5, (Mouse) Q91ZV7
Entrez Gene ID: (Human) 57125, (Mouse) 72324
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support