Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: TRSRKLPHEL CTLIRNVKPE DVKRVASKML RGKPAVAALG DLTDLPTYEH IQTALSSKDG RLPRTYRLFR
Substrate recognition and binding subunit of the essential mitochondrial processing protease (MPP), which is required for maturation of the majority of mitochondrial precursor proteins. Most MPP cleavage sites follow an arginine at position -2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha-MPP; Inactive zinc metalloprotease alpha; inositol polyphosphate-5-phosphatase, 72 kD; mitochondrial matrix processing protease, alpha subunit; mitochondrial processing peptidase alpha subunit, mitochondrial; Mitochondrial-processing peptidase subunit alpha; P-55
Gene Aliases: 1200002L24Rik; 4933435E07Rik; Alpha-MPP; INPP5E; KIAA0123; MPPA; P-55; PMPCA; SCAR2
UniProt ID: (Human) Q10713, (Mouse) Q9DC61
Entrez Gene ID: (Human) 23203, (Mouse) 66865, (Rat) 296588
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support