Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: DLDGTLITT RSGKVFPTGP SDWRILYPEI PRKLRELEAE GYKLVIFTNQ MSIGRGKLPA EEFKAKVEAV VEKLGVPFQV LVATHAGLYR KPVTGMWDHL QEQANDGTPI SIGDSIFVGD AAGRPANWAP GRKKKDFSCA DRLFALNLGL PFATPEEFFL KWPAAGFELP AFDPRTVSRS GPLCLPESRA LLSASPEVVV AVGFPGAGKS TFLKKHLVSA GYVHVNRDTL GSWQRCVTTC ETALKQGKRV AIDNTNPDAA SRARYVQCAR AAGVPCRCFL FTATLEQARH NNRFREMTDS SHIPVSDMVM YGYRKQFEAP TLAEGFSAIL EIPFRLWVEP RLGRLYCQFS EG (171-521 aa encoded by BC033822)
Anti-human Polynucleotide Kinase Mouse Monoclonal Antibody recognizes polynucleotide kinase (PNK), a 57 kDa nuclear enzyme implicated in DNA strand repair processes that occur during DNA replication and recombination, or in response to DNA damaging agents. PNK targets the DNA 5'-hydroxy and 3'-phosphate termini for phosphorylation and dephosphorylation events. During strand breakage in human cells, normal PNK associates with other nuclear proteins to prepare the DNA termini for the enzymatic actions of DNA polymerase-beta and DNA ligase III. PNK is a key functional component in maintaining the genetic stability of human cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Bifunctional polynucleotide phosphatase/kinase; DNA 5'-kinase/3'-phosphatase; Homo sapiens polynucleotide kinase 3'-phosphatase (PNKP); phosphatase/kinase; PKNP; Polynucleotide kinase-3'-phosphatase
Gene Aliases: 1810009G08Rik; AOA4; EIEE10; MCSZ; PNK; PNKP
UniProt ID: (Human) Q96T60
Entrez Gene ID: (Human) 11284, (Mouse) 59047, (Rat) 308576
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support