Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Sequence of this protein is as follows: MALALLEDWC RIMSVDEQKS LMVTGIPADF EEAEIQEVLQ ETLKSLGRYR LLGKIFRKQE NANAVLLELL EDTDVSAIPS EVQGKGGVWK VIFKTPNQDT EFLERLNLFL EKEGQTVSGM FRALGQEGVS PATVPCISPE LLAHLLGQAM AHAPQPLLPM RYRKLRVFSG SAVPAPEEES FEVWLEQATE IVKEWPVTEA EKKRWLAESL RGPALDLMHI VQADNPSISV EECLEAFKQV FGSLESRRTA QVRYLKTYQE EGEKVSAYVL RLETLLRRAV EKRAIPRRIA DQVRLEQVMA GATLNQMLWC RLRELKDQGP PPSFLELMKV IREEEEEEAS FENESIEEPE ERDGYGRWNH EGDD
In patients with cancer, symptoms of limbic and brain-stem dysfunction may result from a para-neoplastic disorder. Two proteins, Ma1 and Ma2, were discovered by presence of autoantibodies against them. Antibodies to Ma1 and Ma2 proteins identify a paraneoplastic disorder that affects the limbic system, brain stem, and cerebellum. These autoantibodies were used to clone Ma2, which codes for a protein (Ma2) that was recognized by serum from the 10 patients, but not by serum from 344 control subjects. Ma2 was selectively expressed by normal brain tissue and by the testicular tumors of the patients. Ma2 shares homology with Ma1, a "brain-testis-cancer" gene related to other para-neoplastic syndromes and tumors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 40 kDa neuronal protein; Onconeuronal antigen Ma2; Paraneoplastic antigen Ma2; Paraneoplastic neuronal antigen MM2; retrotransposon gag domain containing 2
Gene Aliases: KIAA0883; MA2; MM2; PNMA2; RGAG2
UniProt ID: (Human) Q9UL42
Entrez Gene ID: (Human) 10687
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support