Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 3
Sequence of this protein is as follows: MYDAERGWSL SFAGCGFLGF YHVGATRCLS EHAPHLLRDA RMLFGASAGA LHCVGVLSGI PLEQTLQVLS DLVRKARSRN IGIFHPSFNL SKFLRQGLGK CLPANVHQLI SGKIGISLTR VSDGENVLVS DFRSKDEVVD ALVCSCFMPF YSGLIPPSFR GVRYVDGGVS DNVPFIDAKT TITVSPFYGE YDICPKVKST NFLHVDITKL SLRLCTGNLY LLSRAFVPPD LKVLGEICLR GYLDAFRFLE EKGICNRPQP GLKSSSEGMD PEVAMPSWAN MSLDSSPESA ALAVRLEGDE LLDHLRLSIL PWDESILDTL SPRLATALSE EMKDKGGYMS KICNLLPIRI MSYVMLPCTL PVESAIAIVQ RLVTWLPDMP DDVLWLQWVT SQVFTRVLMC LLPASRSQMP VSSQQASPCT PEQDWPCWTP CSPEGCPAET KAEATPRSIL RSSLNFFLGN KVPAGAEGLS TFPSFSLEKS L
The protein encoded by this gene is a triacylglycerol lipase that mediates triacylglycerol hydrolysis in adipocytes. The encoded protein, which appears to be membrane bound, may be involved in the balance of energy usage/storage in adipocytes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1-acylglycerol-3-phosphate O-acyltransferase PNPLA3; acylglycerol O-acyltransferase; Acylglycerol transacylase; Adiponutrin; ADPN; Calcium-independent phospholipase A2-epsilon; iPLA2-epsilon; iPLA2epsilon; Lysophosphatidic acid acyltransferase; patatin-like phospholipase domain containing 3; Patatin-like phospholipase domain-containing protein 3
Gene Aliases: ADPN; C22orf20; iPLA(2)epsilon; PNPLA3
UniProt ID: (Human) Q9NST1, (Mouse) Q91WW7
Entrez Gene ID: (Human) 80339, (Mouse) 116939
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support