Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • POGZ Antibodies

          Abnova

          POGZ Polyclonal Antibody, MaxPab™

          View all (13) POGZ antibodies
          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite POGZ Polyclonal Antibody, MaxPab™

          • Antibody Testing Data (1)
          POGZ Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          POGZ Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          POGZ Antibody (H00023126-B01P) in WB

          Western Blot analysis of POGZ expression in transfected 293T cell line by POGZ MaxPab polyclonal antibody. Lane 1: POGZ transfected lysate(149.27 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          POGZ Antibody in Western Blot (WB)

          Product Details

          H00023126-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Immunocytochemistry (ICC/IF)

          10-100 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          POGZ (NP_997054.1, 1 a.a. approximately 1357 a.a) full-length human protein.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Wet ice

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MADTDLFMEC EEEELEPWQK ISDVIEDSVV EDYNSVDKTT TAGNPLVQQG GQPLILTQNP APGLGTMVTQ PVLRPVQVMQ NANHVTSSPV ASQPIFITTQ GFPVRNVRPV QNAMNQVGIV LNVQQGQTVR PITLVPAPGT QFVKPTVGVP QVFSQMTPVR PGSTMPVRPT TNTFTTVIPA TLTIRSTVPQ SQSQQTKSTP STSTTPTATQ PTSLGQLAVQ SPGQSNQTTN PKLAPSFPSP PAVSIASFVT VKRPGVTGEN SNEVAKLVNT LNTIPSLGQS PGPVVVSNNS SAHGSQRTSG PESSMKVTSS IPVFDLQDGG RKICPRCNAQ FRVTEALRGH MCYCCPEMVE YQKKGKSLDS EPSVPSAAKP PSPEKTAPVA STPSSTPIPA LSPPTKVPEP NENVGDAVQT KLIMLVDDFY YGRDGGKVAQ LTNFPKVATS FRCPHCTKRL KNNIRFMNHM KHHVELDQQN GEVDGHTICQ HCYRQFSTPF QLQCHLENVH SPYESTTKCK ICEWAFESEP LFLQHMKDTH KPGEMPYVCQ VCQYRSSLYS EVDVHFRMIH EDTRHLLCPY CLKVFKNGNA FQQHYMRHQK RNVYHCNKCR LQFLFAKDKI EHKLQHHKTF RKPKQLEGLK PGTKVTIRAS RGQPRTVPVS SNDTPPSALQ EAAPLTSSMD PLPVFLYPPV QRSIQKRAVR KMSVMGRQTC LECSFEIPDF PNHFPTYVHC SLCRYSTCCS RAYANHMINN HVPRKSPKYL ALFKNSVSGI KLACTSCTFV TSVGDAMAKH LVFNPSHRSS SILPRGLTWI AHSRHGQTRD RVHDRNVKNM YPPPSFPTNK AATVKSAGAT PAEPEELLTP LAPALPSPAS TATPPPTPTH PQALALPPLA TEGAECLNVD DQDEGSPVTQ EPELASGGGG SGGVGKKEQL SVKKLRVVLF ALCCNTEQAA EHFRNPQRRI RRWLRRFQAS QGENLEGKYL SFEAEEKLAE WVLTQREQQL PVNEETLFQK ATKIGRSLEG GFKISYEWAV RFMLRHHLTP HARRAVAHTL PKDVAENAGL FIDFVQRQIH NQDLPLSMIV AIDEISLFLD TEVLSSDDRK ENALQTVGTG EPWCDVVLAI LADGTVLPTL VFYRGQMDQP ANMPDSILLE AKESGYSDDE IMELWSTRVW QKHTACQRSK GMLVMDCHRT HLSEEVLAML SASSTLPAVV PAGCSSKIQP LDVCIKRTVK NFLHKKWKEQ AREMADTACD SDVLLQLVLV WLGEVLGVIG DCPELVQRSF LVASVLPGPD GNINSPTRNA DMQEELIASL EEQLKLSGEH SESSTPRPRS SPEETIEPES LHQLFEGESE TESFYGFEEA DLDLMEI

          Target Information

          POGZ appears to be a zinc finger protein containing a transposase domain at the C-terminus. This protein was found to interact with the transcription factor SP1 in a yeast two-hybrid systemThe protein encoded by this gene appears to be a zinc finger protein containing a transposase domain at the C-terminus. This protein was found to interact with the transcription factor SP1 in a yeast two-hybrid system. At least three alternatively spliced transcript variants encoding distinct isoforms have been observed.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Pogo transposable element with ZNF domain; putative protein product of Nbla00003; Suppressor of hairy wing homolog 5; Zinc finger protein 280E; Zinc finger protein 635

          View more View less

          Gene Aliases: KIAA0461; MRD37; Nbla00003; POGZ; SUHW5; WHSUS; ZNF280E; ZNF635; ZNF635m

          View more View less

          UniProt ID: (Human) Q7Z3K3

          View more View less

          Entrez Gene ID: (Human) 23126

          View more View less

          Function(s)
          DNA binding protein binding metal ion binding
          Process(es)
          mitotic sister chromatid cohesion cell division kinetochore assembly
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7fd48b9cb8-69xq6:80/100.66.134.130:80.
          git-commit: fe0b5dd8e7a67b57f27a6e2d26c18f8954d91d59
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0