Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MAPERLRSRA LSAFKLRGLL LRGEAIKYLT EALQSISELE LEDKLEKIIN AVEKQPLSSN MIERSVVEAA VQECSQSVDE TIEHVFNIIG AFDIPRFVYN
DNA replication, recombination and repair, all of which are necessary for genome stability, require the presence of exonucleases. In DNA replication, these enzymes are involved in the processing of Okazaki fragments, whereas in DNA repair, they function to excise damaged DNA fragments and correct recombinational mismatches. Exonucleases involved in these processes include DNA polymerases, including DNA pol delta and epsilon. DNA pol delta consists of two subunits-p125 which interacts directly with the sliding DNA clamp protein PCNA, and p50. DNA pol delta can be regulated by cell cycle proteins. DNA pol epsilon is a multiple subunit enzyme, the catalytic subunit of which is encoded by the POL2 gene. The exact reactions catalyzed by DNA pol delta and epsilon on leading and lagging strands have not yet been elucidated.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA pol epsilon2; DNA pol epsilonB; DNA polymerase epsilon small subunit; DNA polymerase epsilon subunit 2; DNA polymerase epsilon subunit B; DNA polymerase epsilon, subunit 2; DNA polymerase II subunit 2; DNApol epsilon2; DNApol epsilonB; polymerase (DNA directed), epsilon 2 (p59 subunit); polymerase (DNA directed), epsilon 2, accessory subunit; polymerase (DNA) epsilon 2, accessory subunit
Gene Aliases: DPE2; POLE2
UniProt ID: (Human) P56282, (Mouse) O54956
Entrez Gene ID: (Human) 5427, (Rat) 299112, (Mouse) 18974
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support