Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MATGQDRVVA LVDMDCFFVQ VEQRQNPHLR NKPCAVVQYK SWKGGGIIAV SYEARAFGVT RSMWADDAKK LCPDLLLAQV RESRGKANLT KYREASVEVM EIMSRFAVIE RASIDEAYVD LTSAVQERLQ KLQGQPISAD LLPSTYIEGL PQGPTTAEET VQKEGMRKQG LFQWLDSLQI DNLTSPDLQL TVGAVIVEEM RAAIERETGF QCSAGISHNK VLAKLACGLN KPNRQTLVSH GSVPQLFSQM PIRKIRSLGG KLGASVIEIL GIEYMGELTQ FTESQLQSHF GEKNGSWLYA MCRGIEHDPV KPRQLPKTIG CSKNFPGKTA LATREQVQWW LLQLAQELEE RLTKDRNDND RVATQLVVSI RVQGDKRLSS LRRCCALTRY DAHKMSHDAF TVIKNCNTSG IQTEWSPPLT MLFLCATKFS ASAPSSSTDI TSFLSSDPSS LPKVPVTSSE AKTQGSGPAV TATKKATTSL ESFFQKAAER QKVKEASLSS LTAPTQAPMS NSPSKPSLPF QTSQSTGTEP FFKQKSLLLK QKQLNNSSVS SPQQNPWSNC KALPNSLPTE YPGCVPVCEG VSKLEESSKA TPAEMDLAHN SQSMHASSAS KSVLEVTQKA TPNPSLLAAE DQVPCEKCGS LVPVWDMPEH MDYHFALELQ KSFLQPHSSN PQVVSAVSHQ GKRNPKSPLA CTNKRPRPEG MQTLESFFKP LTH
The XPA gene encodes a zinc metalloprotein that preferentially binds to DNA damaged by UV radiation and chemical carcinogens and is required for the incision step during nucleotide excision repair The XPB and XPD genes encode DNA helicases involved in several DNA metabolic pathways, including DNA repair and transcription, and the XPG gene product is an endonuclease that cuts on the 3' side of a DNA lesion during nucleotide excision repair. Molecular defects in the XP variant (POL H) group maintain normal excision repair, yet they result in a substantial reduction in the ability to synthesize intact daughter DNA strands during DNA replication following DNA damage.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA polymerase eta; FLJ16395; FLJ21978; polymerase (DNA directed), eta; polymerase (DNA) eta; RAD30 homolog A; Xeroderma pigmentosum variant type protein; XP V
Gene Aliases: POLH; RAD30; RAD30A; XP-V; XPV
UniProt ID: (Human) Q9Y253
Entrez Gene ID: (Human) 5429
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support