Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • POLR1C Antibodies

          Proteintech

          POLR1C Polyclonal Antibody

          1 Reference
          View all (12) POLR1C antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite POLR1C Polyclonal Antibody

          • Antibody Testing Data (21)
          POLR1C Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          POLR1C Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 21

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          POLR1C Antibody (15923-1-AP) in IHC (P)

          Immunohistochemistry of paraffin-embedded human placenta using 15923-1-AP (POLR1C antibody) at dilution of 1:200 (under 40x lens). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          POLR1C Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          POLR1C Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          POLR1C Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          POLR1C Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          POLR1C Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          POLR1C Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          POLR1C Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          POLR1C Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          POLR1C Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          POLR1C Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          POLR1C Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          POLR1C Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          POLR1C Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          POLR1C Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          POLR1C Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          POLR1C Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          POLR1C Antibody in Western Blot (WB)
          POLR1C Antibody in Western Blot (WB)
          POLR1C Antibody in Western Blot (WB)
          POLR1C Antibody in Western Blot (WB)
          POLR1C Antibody in Western Blot (WB)

          Product Details

          15923-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          -

          Not applicable (N/A)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          POLR1C Fusion Protein Ag8746 (1-346 aa encoded by BC008863)
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.13 mg/mL

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: MAASQAVEE MRSRVVLGEF GVRNVHTTDF PGNYSGYDDA WDQDRFEKNF RVDVVHMDEN SLEFDMVGID AAIANAFRRI LLAEVPTMAV EKVLVYNNTS IVQDEILAHR LGLIPIHADP RLFEYRNQGD EEGTEIDTLQ FRLQVRCTRN PHAAKDSSDP NELYVNHKVY TRHMTWIPLG NQADLFPEGT IRPVHDDILI AQLRPGQEID LLMHCVKGIG KDHAKFSPVA TASYRLLPDI TLLEPVEGEA AEELSRCFSP GVIEVQEVQG KKVARVANPR LDTFSREIFR NEKLKKVVRL ARVRDHYIFS VESTGVLPPD VLVSEAIKVL MGKCRRFLDE LDAVQMD (1-346 aa encoded by BC008863)

          Target Information

          RNA polymerases transcribe nuclear genes for ribosomal RNA, thus representing ribosomal biogenesis. RNA polymerase I (Pol I) is located in the nucleolus and transcribes class I genes, which code for large ribosomal RNA. Different subunits of the Pol I transcription machinery are targets of various physiological stimuli, which suggests that multiple signaling pathways are involved in carrying out Pol I transcription. RPA16, RPA40 and RPA135 are subunits of Pol I that associate with each other at an early stage of RNA Pol I assembly. RPA40 is essential for the function and integrity of the complex and is also an essential subunit of RNA polymerase III (Pol III).

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: AC40; DNA-directed RNA polymerase I subunit C; DNA-directed RNA polymerases I and III 40 kDa polypeptide; DNA-directed RNA polymerases I and III subunit RPAC1; polymerase (RNA) I polypeptide C; polymerase (RNA) I polypeptide C, 30kDa; polymerase (RNA) I subunit C; RNA polymerases I and III subunit AC1; RPA39; RPA40; RPC40

          View more View less

          Gene Aliases: AC40; HLD11; POLR1C; POLR1E; RPA39; RPA40; RPA5; RPAC1; RPC40; TCS3

          View more View less

          UniProt ID: (Human) O15160

          View more View less

          Entrez Gene ID: (Human) 9533

          View more View less

          Function(s)
          RNA polymerase I activity RNA polymerase III activity DNA binding DNA-directed RNA polymerase activity protein binding protein dimerization activity
          Process(es)
          transcription from RNA polymerase I promoter transcription initiation from RNA polymerase I promoter transcription elongation from RNA polymerase I promoter termination of RNA polymerase I transcription transcription from RNA polymerase III promoter positive regulation of type I interferon production positive regulation of gene expression, epigenetic
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0