Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Peptide Sequence: MAASQAVEEM RSRVVLGEFG VRNVHTTDFP GNYSGYDDAW DQDRFEKNFR VDVVHMDENS LEFDMVGIDA AIANAFRRIL LAEVPTMAVE KVLVYNNTSI VQDEILAHRL GLIPIHADPR LFEYRNQGDE EGTEIDTLQF RLQVRCTRNP HAAKDSSDPN ELYVNHKVYT RHMTWIPLGN QADLFPEGTI RPVHDDILIA QLRPGQEIDL LMHCVKGIGK DHAKFSPVAT ASYRLLPDIT LLEPVEGEAA EELSRCFSPG VIEVQEVQGK KVARVANPRL DTFSREIFRN EKLKKVVRLA RVRDHYIFSV ESTGVLPPDV LVSEAIKVLM GKCRRFLDEL DAVQMD
RNA polymerases transcribe nuclear genes for ribosomal RNA, thus representing ribosomal biogenesis. RNA polymerase I (Pol I) is located in the nucleolus and transcribes class I genes, which code for large ribosomal RNA. Different subunits of the Pol I transcription machinery are targets of various physiological stimuli, which suggests that multiple signaling pathways are involved in carrying out Pol I transcription. RPA16, RPA40 and RPA135 are subunits of Pol I that associate with each other at an early stage of RNA Pol I assembly. RPA40 is essential for the function and integrity of the complex and is also an essential subunit of RNA polymerase III (Pol III).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AC40; DNA-directed RNA polymerase I subunit C; DNA-directed RNA polymerases I and III 40 kDa polypeptide; DNA-directed RNA polymerases I and III subunit RPAC1; polymerase (RNA) I polypeptide C; polymerase (RNA) I polypeptide C, 30kDa; polymerase (RNA) I subunit C; RNA polymerases I and III subunit AC1; RPA39; RPA40; RPC40
Gene Aliases: AC40; HLD11; POLR1C; POLR1E; RPA39; RPA40; RPA5; RPAC1; RPC40; TCS3
UniProt ID: (Human) O15160
Entrez Gene ID: (Human) 9533
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support