Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MAEVKVKVQ PPDADPVEIE NRIIELCHQF PHGITDQVIQ NEMPHIEAQQ RAVAINRLLS MGQLDLLRSN TGLLYRIKDS QNAGKMKGSD NQEKLVYQII EDAGNKGIWS RDIRYKSNLP LTEINKILKN LESKKLIKAV KSVAASKKKV YMLYNLQPDR SVTGGAWYSD QDFESEFVEV LNQQCFKFLQ SKAETARESK QNPMIQRNSS FASSHEVWKY ICELGISKVE LSMEDIETIL NTLIYDGKVE MTIIAAKEGT VGSVDGHMKL YRAVNPIIPP TGLVRAPCGL CPVFDDCHEG GEISPSNCIY MTEWLEF (1-316 aa encoded by BC012588)
The human POLR3F is a component of RNA III polymerase. RNA polymerase III transcribes many essential, small, noncoding RNAs, including the 5S rRNAs and tRNAs. While most pol III-transcribed genes are found scattered throughout the linear chromosome maps or in multiple linear clusters, there is increasing evidence that many of these genes prefer to be spatially clustered, often at or near the nucleolus. This association could create an environment that fosters the coregulation of transcription by pol III with transcription of the large ribosomal RNA repeats by RNA polymerase I (pol I) within the nucleolus. Given the high number of pol III-transcribed genes in all eukaryotic genomes, the spatial organization of these genes is likely to affect a large portion of the other genes in a genome. POLR3F has also been recently identified as an HIV dependency factor (HDF), suggesting that POLR3F may be an important drug target in HIV treatment. At least two isoforms of POLR3F are known to exist.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA-directed RNA polymerase III subunit F; DNA-directed RNA polymerase III subunit RPC6; polymerase (RNA) III (DNA directed) polypeptide F, 39 kDa; polymerase (RNA) III subunit F; RNA polymerase III 39 kDa subunit; RNA polymerase III C39 subunit; RNA polymerase III subunit C6; RPC39
Gene Aliases: 2810411G20Rik; 3010019O03Rik; 3110032A07Rik; POLR3F; RPC39; RPC6
UniProt ID: (Human) Q9H1D9, (Mouse) Q921X6
Entrez Gene ID: (Human) 10621, (Mouse) 70408, (Rat) 311487
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support