Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive Samples: SH-SY5Y, Mouse kidney, Mouse heart, Mouse brain, Rat testis
Immunogen sequence: HLLEEDDSLY CISAWNDQGY EHTAEDPALL YRVETMPGLG WVLRRSLYKE ELEPKWPTPE KLWDWDMWMR MPEQRRGREC IIPDVSRSYH FGIVGLNMNG YFHEAYFKKH KFNTVPGVQL RNVDSLKKEA YEVEVHRLLS EAEVLDHSKN PCEDSFLPDT EGHTYVAFIR MEKDDDFTTW TQLAKCLHIW DLDVRGNHRG LWRLFRKKNH FLVVGVPASP YSVKKPPSVT PIFLEPPPKE EGAPGAPEQT
The product of the POMGNT1 gene, protein O-mannose beta-1,2-N-acetylglucosaminyltransferase, participates in O-mannosyl glycan synthesis. POMGnT1 is an N(in)/C(out) (type II) membrane protein localized in the medial-Golgi that initiates the conversion of high mannose N-glycans to complex N-glycans. Specifically, POMGnT1 is a glycosylation enzyme that participates in the synthesis of O-mannosyl glycan, a laminin-binding ligand of alpha-dystroglycan that is rarely synthesized in mammals. Mutations in the POMGNT1 gene cause muscle-eye-brain disease (MEB), an autosomal recessive disorder characterized by congenital muscular dystrophy, ocular abnormalities and lissencephaly.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GnT I.2; MDDGA3; MDDGB3; MDDGC3; O-linked mannose beta1,2-N-acetylglucosaminyltransferase; O-mannosyl N-acetylglucosaminyltransferase; OMGnT1; POMGnT1; protein O-linked mannose beta1,2-N-acetylglucosaminyltransferase; Protein O-linked-mannose beta-1,2-N-acetylglucosaminyltransferase 1; RP11-322N21.3; UDP-GlcNAc:alpha-D-mannoside beta-1,2-N-acetylglucosaminyltransferase I.2
Gene Aliases: 0610016I07Rik; 4930467B06Rik; GnT I.2; gnT-I.2; GNTI.2; LGMD2O; MEB; MGAT1.2; POMGNT1; UNQ746/PRO1475
UniProt ID: (Human) Q8WZA1, (Rat) Q5XIN7, (Mouse) Q91X88
Entrez Gene ID: (Human) 55624, (Rat) 362567, (Mouse) 68273
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support