Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MLWQKPTAP EQAPAPARPY QGVRVKEPVK ELLRRKRGHA SSGAAPAPTA VVLPHQPLAT YTTVGPSCLD MEGSVSAVTE EAALCAGWLS QPTPATLQPL APWTPYTEYV PHEAVSCPYS ADMYVQPVCP SYTVVGPSSV LTYASPPLIT NVTTRSSATP AVGPPLEGPE HQAPLTYFPW PQPLSTLPTS TLQYQPPAPA LPGPQFVQLP ISIPEPVLQD MEDPRRAASS LTIDKLLLEE EDSDAYALNH TLSVEGF (1-256 aa encoded by BC032549)
Transcriptional coactivator that specifically associates with either OCT1 or OCT2. It boosts the OCT1 mediated promoter activity and to a lesser extent, that of OCT2. It has no intrinsic DNA-binding activity. It recognizes the POU domains of OCT1 and OCT2. It is essential for the response of B-cells to antigens and required for the formation of germinal centers.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B-cell-specific coactivator OBF-1; BOB-1; OCA-B; OCT-binding factor 1; POU domain class 2-associating factor 1; POU domain, class 2, associating factor 1
Gene Aliases: BOB1; OBF-1; OBF1; OCAB; POU2AF1
UniProt ID: (Human) Q16633
Entrez Gene ID: (Human) 5450
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support