Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VTLFSAPNYC GEFDNAGAMM SVDETLMCSF QILKPAEKKK PNATRPVTPP
For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles.
Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%
The protein encoded by this gene belongs to the protein phosphatase family, PP1 subfamily. PP1 is an ubiquitous serine/threonine phosphatase that regulates many cellular processes, including cell division. It is expressed in mammalian cells as three closely related isoforms, alpha, beta/delta and gamma, which have distinct localization patterns. This gene encodes the gamma isozyme. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC1675; MGC2870; PP-1G; PP1C gamma 1; PP1C gamma 2; PP1G; PP1gamma; PP2CG; PP2CGAMMA; PPP2CG; protein phosphatase 1, catalytic subunit, gamma isoform; protein phosphatase 1, catalytic subunit, gamma isozyme; Protein phosphatase 1C catalytic subunit; serine/threonine phosphatase 1 gamma; Serine/threonine-protein phosphatase PP1-gamma catalytic subunit
Gene Aliases: dis2m1; PP-1G; PP1; PP1C; PPP1CC; PPP1G
UniProt ID: (Human) P36873, (Mouse) P63087
Entrez Gene ID: (Human) 5501, (Mouse) 19047
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support