Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibody detects endogenous levels of total PPAN.
The protein encoded by this gene is an evolutionarily conserved protein similar to yeast SSF1 as well as to the gene product of the Drosophila gene peter pan (ppan). SSF1 is known to be involved in the second step of mRNA splicing. Both SSF1 and ppan are essential for cell growth and proliferation. Exogenous expression of this gene was reported to reduce the anchorage-independent growth of some tumor cells. Read-through transcription of this gene with P2RY11/P2Y(11), an adjacent downstream gene that encodes an ATP receptor, has been found. These read-through transcripts are ubiquitously present and up-regulated during granulocyte differentiation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Brix domain-containing protein 3; homolog of S. cerevisiae SSF1; Peter Pan homolog; second-step splicing factor 1; Ssf-1; suppressor of sterile four 1; Suppressor of SWI4 1 homolog
Gene Aliases: A230087P06Rik; BXDC3; PPAN; SSF; SSF-1; SSF1; SSF2
UniProt ID: (Human) Q9NQ55, (Mouse) Q91YU8
Entrez Gene ID: (Human) 56342, (Rat) 298699, (Mouse) 235036
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support