Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: ATTKRVLYVG GLAEEVDDKV LHAAFIPFGD ITDIQIPLDY ETEKHRGFAF VEFELAEDAA AAIDNMNESE LFGRTIRVNL AKPMRIKEGS SRPVWSDDD
The protein encoded by this gene is a member of the peptidyl-prolyl cis-trans isomerase (PPIase) family. PPIases catalyze the cis-trans isomerization of proline imidic peptide bonds in oligopeptides and accelerate the folding of proteins. This protein contains a highly conserved cyclophilin (CYP) domain as well as an RNA-binding domain. It was shown to possess PPIase and protein folding activities and also exhibit RNA-binding activity. Three alternatively spliced transcript variants encoding distinct isoforms have been observed.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cyclophilin E; Cyclophilin-33; Peptidyl-prolyl cis-trans isomerase E; peptidylprolyl isomerase E (cyclophilin E); PPIase E; Rotamase E
Gene Aliases: 2010010D16Rik; Ab1-210; CYP-33; CYP33; PPIE
UniProt ID: (Human) Q9UNP9, (Mouse) Q9QZH3
Entrez Gene ID: (Human) 10450, (Rat) 298508, (Mouse) 56031
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support