Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Peptide sequence: SFCSVDPYNP QNFTATIQTA ARIVPEEPSD SEKDLSGKSD LENSSQSGSL
Sequence homology: Cow: 92%; Dog: 91%; Human: 100%
Maintains low levels of EIF2S1 phosphorylation in unstressed cells by promoting its dephosphorylation by PP1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein phosphatase 1 regulatory subunit 15B; protein phosphatase 1, regulatory (inhibitor) subunit 15B
Gene Aliases: CREP; MSSGM2; PPP1R15B
UniProt ID: (Human) Q5SWA1
Entrez Gene ID: (Human) 84919
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support