Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
OriGene
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
In eukaryotes, the phosphorylation and dephosphorylation of proteins on serine and threonine residues is an essential means of regulating a broad range of cellular functions, including division, homeostasis and apoptosis. A group of proteins that are intimately involved in this process are the protein phosphatases. In general, the protein phosphatase 1 (PP1) holoenzyme is a trimeric complex composed of a regulatory subunit, a variable subunit, and a catalytic subunit. Sds22, also known as PPP1R7 (protein phosphatase 1, regulatory (inhibitor) subunit 7), is a 360 amino acid protein that localizes to the nucleus and contains ten LRR (leucine rich) repeats. Expressed in a variety of tissues, Sds22 functions as a regulatory subunit of the PP1 complex, suggesting a role in protein regulation throughout the cell. Multiple isoforms of Sds22 exist due to alternative splicing events.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein phosphatase 1 regulatory subunit 22; Protein phosphatase 1 regulatory subunit 7; protein phosphatase 1, regulatory (inhibitor) subunit 7; protein phosphatase 1, regulatory subunit 7; testis secretory sperm-binding protein Li 210a
Gene Aliases: PPP1R7; SDS22
UniProt ID: (Human) Q15435
Entrez Gene ID: (Human) 5510, (Dog) 100855698
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support