Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MGGEDDELKG LLGLPEEETE LDNLTEFNTA HNKRISTLTI EEGNLDIQRP KRMRKNSRVT FSEDDEIINP EDVDPSVGRF RNMVQTAVVP VKKKRVEGPG SLGLEESGSR RMQNFAFSGG LYGGLPPTHS EAGSQPHGIH GTALIGGLPM PYPNLAPDVD LTPVVPSAVN MNPAPNPAVY NPEAVNEPKK KKYAKEAWPG KKPTPSLLI
This gene, through alternative splicing, encodes three different isoforms. Two of the protein isoforms encoded by this gene are specific inhibitors of type 1 serine/threonine protein phosphatases and can bind but not cleave RNA. The third protein isoform lacks the phosphatase inhibitory function but is a single-strand endoribonuclease comparable to RNase E of E. coli. This isoform requires magnesium for its function and cleaves specific sites in A+U-rich regions of RNA.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: activator of RNA decay; NIPP-1; Nuclear inhibitor of protein phosphatase 1; nuclear inhibitor of protein phosphatase-1 alpha; nuclear inhibitor of protein phosphatase-1 beta; nuclear subunit of PP-1; Protein phosphatase 1 regulatory inhibitor subunit 8; protein phosphatase 1, regulatory (inhibitor) subunit 8; protein phosphatase 1, regulatory subunit 8; RNase E
Gene Aliases: 6330548N22Rik; ARD-1; ARD1; AU044684; NIPP-1; NIPP1; PPP1R8; PRO2047
UniProt ID: (Human) Q12972, (Mouse) Q8R3G1
Entrez Gene ID: (Human) 5511, (Rat) 313030, (Mouse) 100336
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support