Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • PPP2R2B Antibodies

          Invitrogen

          PPP2R2B Polyclonal Antibody

          View all (16) PPP2R2B antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite PPP2R2B Polyclonal Antibody

          • Antibody Testing Data (5)
          PPP2R2B Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          PPP2R2B Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 5

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PPP2R2B Antibody (PA5-59878) in ICC/IF

          Immunofluorescent staining of PPP2R2A/PPP2R2B in human cell line A-431 using a PPP2R2A/PPP2R2B Polyclonal Antibody (Product # PA5-59878) shows localization to cytosol. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PPP2R2B Antibody in Immunocytochemistry (ICC/IF)
          PPP2R2B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PPP2R2B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PPP2R2B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PPP2R2B Antibody in Immunohistochemistry (Paraffin) (IHC (P))

          Product Details

          PA5-59878

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.04-0.4 µg/mL
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:200-1:500
          -

          Immunocytochemistry (ICC/IF)

          0.25-2 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant protein corresponding to Human PPP2R2A/PPP2R2B. Recombinant protein control fragment (Product #RP-104328).

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.1 mg/mL

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.2, with 40% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2645885

          Product Specific Information

          Immunogen sequence: YNVYSTFQSH EPEFDYLKSL EIEEKINKIR WLPQQNA

          Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

          Target Information

          The product of this gene belongs to the phosphatase 2 regulatory subunit B family. Protein phosphatase 2 is one of the four major Ser/Thr phosphatases, and it is implicated in the negative control of cell growth and division. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. The B regulatory subunit might modulate substrate selectivity and catalytic activity. This gene encodes a beta isoform of the regulatory subunit B55 subfamily. Defects in this gene cause autosomal dominant spinocerebellar ataxia 12 (SCA12), a disease caused by degeneration of the cerebellum, sometimes involving the brainstem and spinal cord, and in resulting in poor coordination of speech and body movements. Multiple alternatively spliced variants, which encode different isoforms, have been identified for this gene. The 5' UTR of some of these variants includes a CAG trinucleotide repeat sequence (7-28 copies) that can be expanded to 66-78 copies in cases of SCA12.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: BRbeta B-regulatory subunit of protein phosphatase 2A; PP2A PP2ABBETA; PP2A subunit B isoform B55-beta; PP2A subunit B isoform beta; PP2A subunit B isoform BRB; PP2A subunit B isoform PR55-beta; PP2A subunit B isoform R2-beta; PP2A, subunit B, B-beta isoform; PP2A, subunit B, B55-beta isoform; PP2A, subunit B, BRB isoform; PP2A, subunit B, PR55-beta isoform; PP2A, subunit B, R2-beta isoform; PR2A PR2ABBETA; protein phosphatase 2 (formerly 2A), regulatory subunit B (PR 52), beta isoform; protein phosphatase 2 (formerly 2A), regulatory subunit B, beta isoform; protein phosphatase 2 regulatory subunit B, beta; protein phosphatase 2, regulatory subunit B, beta; serine/threonine protein phosphatase 2A, 55 kDa regulatory subunit B, beta isoform; serine/threonine protein phosphatase 2A, neuronal isoform; Serine/threonine-protein phosphatase 2A 55 kDa regulatory subunit B beta isoform

          View more View less

          Gene Aliases: 2900026H06Rik; 6330404L05Rik; B55BETA; E130009M08Rik; PP2A-PR55B; PP2AB55BETA; PP2ABBETA; PP2APR55B; PP2APR55BETA; PPP2R2B; Pppr2b2; PR2AB55BETA; PR2ABBETA; PR2APR55BETA; PR52B; PR55-BETA; PR55BETA; SCA12

          View more View less

          UniProt ID: (Human) Q00005, (Rat) P36877, (Mouse) Q6ZWR4

          View more View less

          Entrez Gene ID: (Human) 5521, (Rat) 60660, (Mouse) 72930

          View more View less

          Function(s)
          protein binding protein phosphatase type 2A regulator activity protein complex binding
          Process(es)
          apoptotic process regulation of protein phosphatase type 2A activity mitochondrial fission protein targeting to mitochondrion signal transduction spermatid development brain development neuron differentiation positive regulation of neuron apoptotic process mitochondrial fragmentation involved in apoptotic process
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0