Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MDVERLQEA LKDFEKRGKK EVCPVLDQFL CHVAKTGETI IPFTIQRLCE LLTDPRRNYT GTDKFLRGVE KNVMVVSCVY PSSEKNNSNS LNRMNGVMFP GNSPSYTERS NINGPGTPRP LNRPKVSLSA PMTTNGLPES TDSKEANLQQ NEEKNHSDSS TSESEVSSVS PLKNKHPDED AVEAEGHEVK RLRFDKEGEV RETASQ (1-205 aa encoded by B C110889)
The protein encoded by this gene is a regulatory subunit of the serine/threonine-protein phosphatase 4 complex. In addition to being required for efficient DNA double strand break repair, this complex plays a role in organization of microtubules at centrosomes and processing of spliceosomal snRNPs. Several transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC131930; protein phosphatase 4, regulatory subunit 2; Serine/threonine-protein phosphatase 4 regulatory subunit 2
Gene Aliases: PP4R2; PPP4R2; SBBI57
UniProt ID: (Human) Q9NY27
Entrez Gene ID: (Human) 151987
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support