Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAMAEGERTE CAEPPRDEPP ADGALKRAEE LKTQANDYFK AKDYENAIKF YSQAIELNPS NAIYYGNRSL AYLRTECYGY ALGDATRAIE LDKKYIKGYY RRAASNMALG KFRAALRDYE TVVKVKPHDK DAKMKYQECN KIVKQKAFER AIAGDEHKRS VVDSLDIESM TIEDEYSGPK LEDGKVTISF MKELMQWYKD QKKLHRKCAY QILVQVKEVL SKLSTLVETT LKETEKITVC GDTHGQFYDL LNIFELNGLP SETNPYIFNG DFVDRGSFSV EVILTLFGFK LLYPDHFHLL RGNHETDNMN QIYGFEGEVK AKYTAQMYEL FSEVFEWLPL AQCINGKVLI MHGGLFSEDG VTLDDIRKIE RNRQPPDSGP MCDLLWSDPQ PQNGRSISKR GVSCQFGPDV TKAFLEENNL DYIIRSHEVK AEGYEVAHGG RCVTVFSAPN YCDQMGNKAS YIHLQGSDLR PQFHQFTAVP HPNVKPMAYA NTLLQLGMM
This gene encodes a serine/threonine phosphatase which is a member of the protein phosphatase catalytic subunit family. Proteins in this family participate in pathways regulated by reversible phosphorylation at serine and threonine residues; many of these pathways are involved in the regulation of cell growth and differentiation. The product of this gene has been shown to participate in signaling pathways in response to hormones or cellular stress, and elevated levels of this protein may be associated with breast cancer development. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ36922; FLJ55954; OTTHUMP00000165834; PP-T; PP5; protein phosphatase 5, catalytic subunit; Protein phosphatase T; Serine/threonine-protein phosphatase 5
Gene Aliases: PP5; PPP5; PPP5C; PPT
UniProt ID: (Human) P53041
Entrez Gene ID: (Human) 5536
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support