Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MPLPVALQTR LAKRGILKHL EPEPEEEIIA EDYDDDPVDY EATRLEGLPP SWYKVFDPSC GLPYYWNADT DLVSWLSPHD PNSVVTKSAK KLRSSNADAE EKLDRSHDKS DRGHDKSDRS HEKLDRGHDK SDRGHDKSDR DRERGYDKVD RERERDRERD RDRGYDKADR EEGKERRHHR REELAPYPKS KKAVSRKDEE LDPMDPSSYS DAPRGTWSTG LPKRNEAKTG ADTTAAGPLF QQRPYPSPGA VLRANAEASR TKQQD
PQBP1 is a nuclear polyglutamine-binding protein that contains a WW domain. Mutations in this gene are associated with X-linked mental retardation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 38 kDa nuclear protein containing a WW domain; Npw38; Polyglutamine tract-binding protein 1; Polyglutamine-binding protein 1; PQBP-1
Gene Aliases: JM26; MRX2; MRX55; MRXS3; MRXS8; NPW38; PQBP1; RENS1; SHS
UniProt ID: (Human) O60828
Entrez Gene ID: (Human) 10084
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support